Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant Kallikrein 10 (from Cells)
Human recombinant Kallikrein 10 (from Cells)
Catalog # PRSI91-331
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Kallikrein 10 (from Cells)
Catalog # PRSI91-331
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    NES1|Protease Serine-like 1|Kallikrein-10|Normal Epithelial Cell-Specific 1|PRSSL1|KLK10
  • Protein/peptide name:
    Kallikrein 10
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    27.98 kD
  • Sequence:
    Ala31-Asn276
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Kallikreins are a subgroup of Serine Proteases having diverse physiological functions. Growing evidence suggests that many Kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen Kallikrein subfamily members located in a cluster on chromosome 19. Its encoded protein is secreted and may play a role in suppression of tumorigenesis in breast and prostate cancers. Alternate splicing of this gene results in multiple transcript variants encoding the same protein.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag