Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:PPIase E|PPIE|Rotamase E|CYP33|Cyclophilin E|Cyclophilin-33|Peptidyl-Prolyl Cis-Trans Isomerase E
- Protein/peptide name:Cyclophilin E
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:35.6 kD
- Sequence:Met1-Val301
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Peptidyl-prolyl cis-trans isomerase E, also known as Cyclophilin E, Cyclophilin-33, Rotamase E, CYP33, PPIE, is an enzyme which belongs to the cyclophilin-type PPIase family of PPIase E subfamily. PPIE found in all the examined tissues including heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas. PPIE contains one PPIase cyclophilin-type domain and one RRM (RNA recognition motif) domain. PPIE accelerates the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. PPIE combines RNA-binding and PPIase activities. It may be involved in muscle- and brain-specific processes and pre-mRNA splici
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag