Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant Annexin A6 (from E. coli)
Human recombinant Annexin A6 (from E. coli)
Catalog # PRSI91-187
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Annexin A6 (from E. coli)
Catalog # PRSI91-187
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Protein III|Lipocortin VI|ANXA6|ANX6|67 kDa Calelectrin|CPB-II|Annexin-6|p68|Calphobindin-II|Annexin A6|p70|Chromobindin-20|Annexin VI
  • Protein/peptide name:
    Annexin A6
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    76.9 kD
  • Sequence:
    Ala2-Asp673
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Annexin A6 (ANAX6) belongs to a family of calcium-dependent membrane and phospholipid binding proteins. Annexin A6 is a secreted protein and locates on the cell surface. Annexin A6 contains 8 annexin repeats separated by linking sequences of variable lengths. A pair of annexin repeats may form one binding site for calcium and phospholipid. ANXA6 is involved in the regulation of the release of Ca2+ from intracellular stores and may be associated with CD21. In addition, ANXA6 has been implicated in mediating the endosome aggregation and vesicle fusion in secreting epithelia during exocytosis.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag