Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:PPM1C|Protein Phosphatase 1G|Protein Phosphatase 2C Isoform Gamma|PPM1G|Protein Phosphatase Magnesium-Dependent 1 Gamma|Protein Phosphatase 1C|PP2C-Gamma
- Protein/peptide name:PP1MG
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:26.15 kD
- Sequence:Met317-Asp546
- Formulation:Supplied as a 0.2 um filtered solution of 25mM TrisHCl, 1mM DTT, 1mM EDTA, 2mM β-ME, 20% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Protein Phosphatase 1G (PP1MG) is a cytoplasmic protein that belongs to the PP2C family. PPM1G is widely expressed; it is abundant in testis, skeletal muscle, and heart. PPM1G is a negative regulator of cell stress response pathways. PPM1G is responsible for the dephosphorylation of Pre-mRNA splicing factors, an important factor for the formation of functional spliceosome. PPM1G also plays a role in regulating cell cycle progression.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag