Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-GST tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:LACTB2|Beta-Lactamase-like Protein 2
- Protein/peptide name:LACTB2
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:59.24 kD
- Sequence:Met1-Leu288
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM EDTA, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:For Biological assays
beta -Lactamase-like Protein 2 (LACTB2) is a number of the metallo-beta-lactamase superfamily.LACTB2 also belongs to the Glyoxalase II family. LACTB2 is 288 amino acids long with 8 zinc-binding domains. The LACTB2 gene is expressed at high levels and annotates structural defects or features in 4 cDNA clones. LACTB2 proteins are expected to have hydrolase activity and metal ion-binding functions. LACTB2 protein is found to localize in mitochondrion. Other functions of LACTB2 is yet unknown.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag