Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:MIP-4-Alpha|Macrophage Inflammatory Protein 4-Alpha|TSC-1|Small-Inducible Cytokine A26|Thymic Stroma Chemokine-1|C-C Motif Chemokine 26|Eotaxin-3|SCYA26|CC Chemokine IMAC|CCL26
- Protein/peptide name:CCL26
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:8.21 kD
- Sequence:Ser27-Leu94
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Chemokine (C C Motif) Ligand 26 (CCL26) is a novel small cytokine belonging to the CC chemokine family, which involved in immunoregulatory and inflammatory processes. CCL26 is expressed constitutively in thymus, but only transiently in phytohemagglutinin-stimulated peripheral blood mononuclear cells. It specifically binds and induces chemotaxis in T cells and elicits its effects by interacting with the chemokine receptor CCR4. Eotaxin-3/CCL26, along with Eotaxin-1 and Eotaxin-2, selectively activates the CC chemokine receptor 3 (CCR3). The Eotaxin-3-CCR3 interaction may play an important role in allergic diseases such as atopic dermatitis and bronchial asthma. The full-length cDNA for Eotaxin-3 encodes a protein of 94 amino acids with a putative signal peptide of either 23 or 26 amino acid residues. Both the 71 and 68 amino acid residue variants of recombinant Eotaxin-3 demonstrate equal potency in inducing chemotaxis of a human CCR3-transfected cell line. Unlike most other CC chemokines, Eotaxin-3 maps to human chromosome 7q11.2, within 40 kilobases of the Eotaxin-2 loci. Eotaxin-3 and Eotaxin-2 are unique in that they are the only chemokines identified to date that map to chromosome 7.
- This recombinant protein can be used for biological assays. For research use only.