Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant Dr51 (from HEK293 cells)
Human recombinant Dr51 (from HEK293 cells)
  PRSI96-749
 :  
undefined
Human recombinant Dr51 (from HEK293 cells)
  PRSI96-749
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its binding ability in a functional ELISA. Immobilized human TNFRSF10B at 10μg/ml ( 100μl/well ) can bind biotinylated TNFSF10 with a linear range of 0. 375 - 10 ng/mL.
  • Protein synonyms:
    TRAILR2|TRAIL-R2|CD262|TRICK2|KILLER|TRICKB|ZTNFR9|TNFRSF10B|DR5
  • Protein/peptide name:
    Dr51
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    15.1 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

 

 

Tumor necrosis factor receptor superfamily member 10B (TNFRSF10B) is also known as TNF-related apoptosis-inducing ligand receptor 2 (TRAILR2), Death receptor 5 (DR5), CD262, KILLER, is a member of the TNF-receptor superfamily, and contains an intracellular death domain. TNFRSF10B / DR-5 is widely expressed in adult and fetal tissues; very highly expressed in tumor cell lines. TRAILR2 / CD262 / DR5 is the receptor for the cytotoxic ligand TNFSF10/TRAIL. The adapter molecule FADD (a death domain containing adaptor protein) of TRAIL-R2 / TNFRSF10B recruits caspase-8 to the activated receptor.

The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. CD262 / DR5 Promotes the activation of NF-kappa-B. DR5 is essential for ER stress-induced apoptosis and is regulated by p53/TP53.