Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:200 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Protein synonyms:L-31|Laminin-binding protein|Galectin-3|IgE-binding protein|LGALS3|GALBP|CBP 35|Lectin L-29|Mac-2 antigen|35 kDa lectin|Gal-3|MAC2
- Protein/peptide name:Galectin 3
- Purity:>92% as determined by SEC-HPLC and SDS-PAGE.
- Molecular weight:27 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot
Galectin-3 (Gal-3) is also known as LGALS3, 35 kDa lectin, Carbohydrate-binding protein 35 (CBP 35), Galactose-specific lectin 3, Galactoside-binding protein (GALBP), IgE-binding protein, Laminin-binding protein, Mac-2 antigen, Lectin L-29. Galectin-3 is a member of the lectin family. LGALS3 / Galectin-3 is expressed in the nucleus, cytoplasm, mitochondrion, cell surface, and extracellular space. LGALS3 / Galectin-3 has been shown to be involved in the following biological processes: cell adhesion, cell activation and chemoattraction, cell growth and differentiation, cell cycle, and apoptosis.