Order Entry
Northern Ireland
ContactUsLinkComponent
Human recombinant PDGF-BB (from E. coli)
Human recombinant PDGF-BB (from E. coli)
Catalog # PRSI40-607
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant PDGF-BB (from E. coli)
Catalog # PRSI40-607
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological activity:
    Determined by the dose-dependent stimulation of the proliferation of Balb/c 3T3 cells. The expected ED50 for this effect is 1.0-3.0 ng/mL.
  • Protein synonyms:
    SIS|PDGF subunit B|PDGF2|c-sis|SSV|Platelet-derived growth factor subunit B|PDGF-2|IBGC5
  • Protein/peptide name:
    PDGF-BB
  • Purity:
    >98%
  • Formulation:
    Lyophilized

Specifications

About this item

PDGFs are disulfide-linked dimers consisting of two 12.0-13.5 kDa polypeptide chains, designated PDGF-A and PDGF-B chains. The three naturally occurring PDGFs; PDGF-AA, PDGF-BB and PDGF-AB, are potent mitogens for a variety of cell types including smooth muscle cells, connective tissue cells, bone and cartilage cells, and some blood cells. The PDGFs are stored in platelet α-granules and are released upon platelet activation. The PDGFs are involved in a number of biological processes, including hyperplasia, chemotaxis, embryonic neuron development, and respiratory tubule epithelial cell development. Two distinct signaling receptors used by PDGFs have been identified and named PDGFR-α and PDGFR-β. PDGFR-α is high-affinity receptor for each of the three PDGF forms. On the other hand, PDGFR-β interacts with only PDGF-BB and PDGF-AB. Recombinant human PDGF-BB is a 24.3 kDa disulfide-linked homodimer of two B chains (218 total amino acids). Recombinant murine PDGF-BB is a 24.4 kDa disulfide-linked homodimer of two B chains (218 total amino acids).

  • Store in working aliquots at -20°C. Avoid freeze/thaw cycles.