Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Human LRRTM1 peptide

Human LRRTM1 peptide

50 µG
Catalog # PRSI6167P
Supplier:   ProSci Inc.
Human LRRTM1 peptide
Catalog # PRSI6167P
Supplier:  ProSci Inc.
CAS Number:  
Human LRRTM1 peptide
Human LRRTM1 peptide

50 µG

CAS Number:  
Price
£117.00
Log in for availability
1 * 50 µG
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

LRRTM1 peptide is used for blocking the activity of LRRTM1 antibody.

Documents

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    PRSI6167P
  • Protein/peptide name:
    LRRTM1 Peptide
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    17 amino acid peptide near the carboxy terminus of human LRRTM1
  • Tested applications:
    Blocking
  • Species:
    Human

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.