Human JMJD2A peptide (14 aa in the center)
Catalog # PRSI5373P
Supplier: ProSci Inc.
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:14 aa in the center
- Protein/peptide name:JMJD2A Peptide (14 aa in the center)
- Tested applications:Blocking
Specifications
About this item
JMJD2A peptide is used for blocking the activity of JMJD2A antibody.