Order Entry
Northern Ireland
ContactUsLinkComponent
Human CTRP1 peptide (16 aa in the center)
Human CTRP1 peptide (16 aa in the center)
Catalog # PRSI3555P
Supplier:  ProSci Inc.
CAS Number:  
Human CTRP1 peptide (16 aa in the center)
Catalog # PRSI3555P
Supplier:  ProSci Inc.
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    16 aa in the center
  • Protein/peptide name:
    CTRP1 Peptide (16 aa in the center)
  • Tested applications:
    Blocking

Specifications

About this item

CTRP1 peptide is used for blocking the activity of CTRP1 antibody.