Human CCR8 peptide (17 aa near the extracellular loop domain)
PRSI2097P
:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Pk:50 µG
- Species:Human
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:17 aa near the extracellular loop domain
- Protein/peptide name:CCR8 Peptide (17 aa near the extracellular loop domain)
- Tested applications:Blocking
CCR8 peptide is used for blocking the activity of CCR8 antibody.