Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen symbol:TNFSF13 / ADIPOQ
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Form:Liquid
- Immunogen:APRIL (monoclonal Sacha-2) antibody was raised against recombinant human APRIL (aa. 105-250).
- Tested applications:Fluorescence-Activated Cell Sorting
- Size:100 µg
- Pk:100 µG
Specifications
About this item
APRIL antibody can be used to recognize human and mouse APRIL by ELISA (capture), Flow Cytometry, and Immunocytochemistry. Method: HEK-293T cells expressing surface membrane associated human APRIL (aa 92-233) or a mock control were injected in chicken liver and frozen in OCT. 8 μm sections of injected frozen liver were fixed in acetone, washed in PBS and incubated with Sacha-2 at 10 μg/mL for 1hr after blocking in PBS/1%BSA. Human APRIL was revealed with 3-amino-9-Ethyl-carbazole (AEC) following 45 min. incubation with goat anti-rat HRP. Method: HEK 293T cells (5 x 105) were mock transfected (thin line) or transfected with an expression plasmid enabling surface expression of mouse APRIL (thick line). Cells were incubated on ice for 30 min in 50 μl FACS buffer (PBS, 5% Fetal calf serum, 0.02% sodium azide) containing 1 μg/mL of Sacha-2 antibody. After washing in FACS buffer, PE-conjugated antibody to rat IgG was added. Cells were incubated on ice for 30 min, washed and then analyzed by flow cytometry. This product is for research use only.
APRIL (monoclonal Sacha-2) antibody was purified from concentrated hybridoma tissue culture supernatant. APRIL (monoclonal Sacha-2) antibody was purified to 95% in SDS-PAGE.
PBS containing 0.02% sodium azide
APRIL antibody can be stored at 4°C, stable for one year.
Type:
Antigen: TNFSF13 / ADIPOQ
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse