Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antigen name:Amyloid beta (A4) Precursor Protein
- Antigen symbol:APP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
- Concentration:1 mg/mL
- Immunogen:APP (Ab-668) antibody was raised against a peptide sequence around aa.666~670 (A-V-T-P-E ) derived from Human APP.
- Size:100 µg
- Pk:100 µG
APP encodes a cell surface receptor and transmembrane precursor protein that is cleaved by secretases to form a number of peptides. Some of these peptides are secreted and can bind to the acetyltransferase complex APBB1/TIP60 to promote transcriptional activation, while others form the protein basis of the amyloid plaques found in the brains of patients with Alzheimer disease. Mutations in this gene have been implicated in autosomal dominant Alzheimer disease and cerebroarterial amyloidosis (cerebral amyloid angiopathy). Multiple transcript variants encoding several different isoforms have been found for this gene.
Western Blot: 1:500~1:1000, Immunofluorescence: 1:100~1:200
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat