Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen name:Transient receptor potential cation channel subfamily V member 4
- Antigen symbol:TRPV4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TrpV4|SMAL|VRL2|SSQTL1|TRP12|VROAC|CMT2C|Transient receptor potential cation channel subfamily V member 4|OTRPC4|HMSN2C|SPSMA|Osm-9-like TRP channel 4|TRPV4
- Concentration:1 mg/mL
- Immunogen:TRPV4 antibody was raised against an 18 amino acid peptide near the center of human TRPV4.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The transient receptor potential (TRP) protein family consists of a diverse group of cation channels functioning in a variety of homeostatic and regulatory pathways. Four subfamilies exist, based on channel domain homology: C type (canonical), V type (vanilloid receptor related), M type (melastatin related) and P type (PKD). TRPV4, belongs to the V type subfamily and plays a role in systemic osmoregulation. TRPV4 is a calcium channel multi-pass membrane protein activated by various stimuli, including thermal stress, fatty acid metabolites and hypotonicity. TRPV4 is highly expressed in lung and kidney and widely expressed in brain. It plays an important role in regulating neural excitability.
TRPV4 antibody is affinity chromatography purified via peptide column.
TRPV4 antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRPV4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human