Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen name:Platelet-derived Growth Factor Beta
- Antigen symbol:PDGFB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:SIS|PDGF subunit B|PDGF2|c-sis|SSV|PDGF-B|Platelet-derived growth factor subunit B|PDGF-2|IBGC5
- Concentration:1 mg/mL
- Immunogen:Rabbit polyclonal PDGF-B antibody was raised against a 19 amino acid peptide near the carboxy terminus of human PDGF-B.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
PDGF-B Antibody: The platelet-derived growth factor beta (PDGF-B) is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB). Mutations in this gene are associated with meningioma , and reciprocal translocations between chromosomes 22 and 7, at sites where the PDGF-B gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans.
PDGF-B Antibody is affinity chromatography purified via peptide column.
PDGF-B Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PDGFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat