Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antigen name:HLA class II histocompatibility antigen, DP(W2) beta chain
- Antigen symbol:HLA-DPB1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:DPB1|HLA-DP|HLA-DPB|HLA-DP1B
- Immunogen:Purified (With Antibiotics) class II from JY cell line.
- Tested applications:Fluorescence-Activated Cell Sorting
- Size:50 µg
- Pk:50 µG
MHC Class II Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 μg/mL.
Protein A Column
50 mM sodium phosphate, pH 7.5, 100 mM potassium chloride, 150 mM NaCl, 0.5 mg/ml Gentamicin sulfate
MHC Class II Monoclonal antibody may be stored at Store at +4°C. Do not freeze.
Type:
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human