Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen name:EPH Receptor B2
- Antigen symbol:EPHB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish,Xenopus,Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:EK5|Hek5|CAPB|ERK|EPHT3|PCBC|Tyro5|DRT
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho-Tyr331 of chicken EphrinB. Note: Chicken Tyr331 is the homolog of mouse and rat Tyr343, human Tyr344 and also Xenopus Tyr324.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
EphrinB proteins are thought to play key roles in cellular functions as diverse as neuronal migration and blood vessel development (Flanagan and Vanderhaeghen, 1998; Dufour et al., 2003; Oike et al., 2002). EphrinB molecules expressed at the membrane surface bind to the EphB family receptors on target cells during cell-to cell contact. This interaction leads to cell signaling in the target cell but also generates a reverse signal in the cell expressing EphrinB on its surface. This reverse signaling event is thought to be critical for vessel maturation and neuronal development. Importantly, tyrosine phosphorylation of EphrinB is thought to be a critical component of this reverse signaling event (Palmer et al., 2002). Recent work demonstrated that Tyr331 of EphrinB was phosphorylated in HEK293 cells after stimulation by the soluble EphB2 receptor tyrosine kinase (Kalo et al., 2001).
The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with bovine, chicken, human, mouse, Xenopus and zebra fish based on the fact that these species have 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EPHB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Bovine, Chicken, Xenopus, Zebrafish