Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-IRAK4 Rabbit Polyclonal Antibody
Anti-IRAK4 Rabbit Polyclonal Antibody
Catalog # PRSI49-298
Supplier:  ProSci Inc.
undefined
Anti-IRAK4 Rabbit Polyclonal Antibody
Catalog # PRSI49-298
Supplier:  ProSci Inc.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen symbol:
    IRAK4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Immunogen:
    IRAK4 antibody was raised against amino acids 38 - 54 of IRAK4 (Mouse).
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

IL-1 receptor-associated kinases (IRAKs) are important mediators in the signal transduction of Toll/IL-1 receptor (TIR) family members. The cytoplasmic domains of TIR proteins interact with the adapter protein, MyD88. MyD88 then recruits IRAKs (IRAK-1, -2, and M), which in turn interact with other adapter molecules, such as TRAF6 to activate NF-κB and MAPK pathways. Recently, a new member of this family, IRAK-4 has been identified. IRAK-4 may act as an upstream activator of IRAK-1. IRAK-4 is important for LPS activation of TLRS. Mice lacking IRAK-4 are resistant to lethal doses of LPS and are also severely impaired in their responses to viral and bacterial challenges (4, 5). IRAK4, an IRAK type protein kinase, is a signal transducer for the immune response Toll-like receptor and interleukin-1 (IL-1) receptor signaling cascades. IRAK4 has auto- and cross-phosphorylation kinase activity and has been shown to phosphorylate and activate IRAK1. Additionally, IRAK4 interacts with IRAK1 and TRAF6 in an IL-1-dependent manner, and overexpression of IRAK4 can activate NF-kappaB as well as mitogen-activated protein (MAP) kinase pathways. IRAK4 (-/-) mice are resistant to a lethal dose of lipopolysaccharide and are severely impaired in their responses to viral and bacterial challenges. At least two mRNA transcripts have been reported: 3.0- and 4.4-kb. IRAK4, also referrred to as NY-REN-64, was first identified as an anitgen in a screen of renal tumors.

IRAK4 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 - 20 μg/mL.

Protein G Column

PBS, 0.05% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IRAK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse