Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ITGA4 Rabbit Polyclonal Antibody
Anti-ITGA4 Rabbit Polyclonal Antibody
Catalog # PRSI48-389
Supplier:  ProSci Inc.
undefined
Anti-ITGA4 Rabbit Polyclonal Antibody
Catalog # PRSI48-389
Supplier:  ProSci Inc.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antigen name:
    Integrin Alpha 4
  • Antigen symbol:
    ITGA4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Hamster,
    Bat,
    Pig,
    Rabbit,
    Monkey,
    Gibbon,
    Gorilla,
    Human,
    Dog,
    Rat,
    Mouse
  • Form:
    Liquid
  • Antigen synonyms:
    CD49D|CD49 antigen-like family member D|IA4|Integrin alpha-4|Integrin alpha 4
  • Immunogen:
    Integrin alpha 4 antibody was raised against a peptide located in the C-Terminal domain of Integrin alpha 4 (Human).
  • Tested applications:
    Immunoaffinity Chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

ITGA4 encodes integrin alpha chain 4. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. Unlike other integrin alpha chains, alpha 4 neither contains an I-domain, nor undergoes disulfide-linked cleavage. Although alpha 4 is variably cleaved to yield two unlinked chains, this cleavage is unnecessary for integrin assembly and function. Alpha 4 combines with beta 1 (ITGB1) on T-cells to form the integrin very late (activation) antigen 4 (VLA-4) that can bind to the extracellular matrix molecules fibronectin or thrombospondin, and is also a ligand for the cell surface molecule vascular cell adhesion molecule 1 (VCAM-1). In addition, alpha 4 combines with beta 7 to form the lymphocyte homing receptor known as LPAM-1 (lymphocyte Peyer Patch adhesion molecule 1). Integrins are also known to participate in cell-surface mediated signalling.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ITGA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Dog, Gibbon, Gorilla, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat