Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen name:Prokineticin Receptor 2
- Antigen symbol:PROKR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:dJ680N4.3|prokineticin receptor 2|PROKR2|GPRg2|KAL3|PKR2|G protein-coupled receptor 73-like 1|GPR73L1|GPR73b|OTTHUMP00000030192
- Concentration:500 ug/mL
- Immunogen:PROKR2 antibody was raised against an 11 amino acid synthetic peptide near the internal region of PROKR2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
PROKR2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
PROKR2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PROKR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat