Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antigen name:deafness, autosomal dominant 5
- Antigen symbol:DFNA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Inversely correlated with estrogen receptor expression 1|Non-syndromic hearing impairment protein 5|ICERE-1
- Immunogen:A synthetic peptide corresponding a region of human DFNA5.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Hearing impairment is a heterogeneous condition with over 40 loci described. DFNA5 is expressed in fetal cochlea, however, its function is not known. Nonsyndromic hearing impairment is associated with a mutation in its gene.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: DFNA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human