Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ATRAID Rabbit Polyclonal Antibody
Anti-ATRAID Rabbit Polyclonal Antibody
Catalog # PRSI25-005
Supplier:  ProSci Inc.
undefined
Anti-ATRAID Rabbit Polyclonal Antibody
Catalog # PRSI25-005
Supplier:  ProSci Inc.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antigen name:
    All-trans Retinoic Acid-induced differentiation Factor
  • Antigen symbol:
    ATRAID
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    ATRAID
  • Immunogen:
    A synthetic peptide corresponding a region of human C2orf28.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

The exact function of C2orf28 remains unknown.This gene is thought to be involved in apoptosis, and may also be involved in hematopoietic development and differentiation. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.

C2orf28 antibody can be used for detection of C2orf28 by ELISA at 1:62500. C2orf28 antibody can be used for detection of C2orf28 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ATRAID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human