Spill trays, EcoPolyBlend™
Eco-friendly spill trays are made from 100% post-industrial recycled PE will not rust or corrode.
- Lightweight, low profile design for small-scale spill control
- Raised rib bottom adds rigidity and keeps contents out of spills
- Rated for service temperatures between –34 to +49 °C
Perfect for pouring and filling applications. Ideal for containing leaks from small equipment or batteries.
Frequently Bought Together