Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ID2 Rabbit Polyclonal Antibody
Anti-ID2 Rabbit Polyclonal Antibody
  BSBTA00417-1
Anti-ID2 Rabbit Polyclonal Antibody
  BSBTA00417-1

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    DNA-binding protein inhibitor ID-2
  • Antigen symbol:
    ID2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for DNA-binding protein inhibitor ID-2(ID2) detection. Tested with WB in Human;Rat.

Type: Primary
Antigen: ID2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat