Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-HBA1|HBA2 Rabbit Polyclonal Antibody
Anti-HBA1|HBA2 Rabbit Polyclonal Antibody
Catalog # BSBTA00233-1
Supplier:  Boster Bio
Anti-HBA1|HBA2 Rabbit Polyclonal Antibody
Catalog # BSBTA00233-1
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Hemoglobin subunit alpha
  • Antigen symbol:
    HBA1|HBA2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Hemoglobin subunit alpha(HBA1|HBA2) detection. Tested with WB, IHC-P in Human;Rat.

Type: Primary
Antigen: HBA1|HBA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Rat