Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-DVL3 Rabbit Polyclonal Antibody
Anti-DVL3 Rabbit Polyclonal Antibody
Catalog # BSBTA03577
Supplier:  Boster Bio
Anti-DVL3 Rabbit Polyclonal Antibody
Catalog # BSBTA03577
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Segment polarity protein dishevelled homolog DVL-3
  • Antigen symbol:
    DVL3
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Segment polarity protein dishevelled homolog DVL-3(DVL3) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: DVL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat