ContactUsLinkComponent
Quantus® AreaDress, Cleanroom Trousers
undefined
Quantus® AreaDress, Cleanroom Trousers
  • Catalog No:
  • 113-9189
  • 113-9191
  • 113-9192
  • 113-9188
  • 113-9187
  • 113-9186
  • 113-9190
  • Model:
  • AreaDress
  • AreaDress
  • AreaDress
  • AreaDress
  • AreaDress
  • AreaDress
  • AreaDress
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Fabric:
  • Polyester fabric with conductive yarn and a PTFE finish
  • Polyester fabric with conductive yarn and a PTFE finish
  • Polyester fabric with conductive yarn and a PTFE finish
  • Polyester fabric with conductive yarn and a PTFE finish
  • Polyester fabric with conductive yarn and a PTFE finish
  • Polyester fabric with conductive yarn and a PTFE finish
  • Polyester fabric with conductive yarn and a PTFE finish
  • Weight:
  • 116 g/m²
  • 116 g/m²
  • 116 g/m²
  • 116 g/m²
  • 116 g/m²
  • 116 g/m²
  • 116 g/m²
Frequently Bought Together