Sunset yellow FCF
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Accessories for AS 200, AS 300 and AS 400
: Retsch GmbH
Riboflavine (vitamin B2) ≥98.0% USP
: Spectrum Chemical