Anti-IgG Rabbit Polyclonal Antibody (Biotin)
30-8077-M002
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Epitope:Heavy and light chain
- Cross adsorption:Yes
- Storage temperature:Store at 2-8 °C
- Pk:2 mg
Rabbit anti-Mouse IgG (H&L) - Affinity Pure, Biotin Conjugate, min x w/human serum proteins
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Rabbit
Isotype:
Reactivity: Mouse