Anti-IgM Goat Polyclonal Antibody
70-8028-M002
- Antibody type:Secondary
- Antigen name:Immunoglobulin M mu
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:µ chain
- Storage temperature:Store at 2-8 °C
- Pk:2 mg
Goat anti-Human IgM (mu chain) - Affinity Pure
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human