Instant sealing pouches, Chex-All® II
Supplier: PROPPER MFG
Certificates
About this item
These instant, self sealing sterilisation pouches are used in steam autoclaves or ethylene oxide sterilisers. This provides a barrier to maintain sterility after the sterilisation process.
- Triple-sealed paper/plastic film provides excellent sterility protection
- In steam autoclaves the yellow indicators turn dark
- In ethylene oxide sterilizers the red indicators change to green
- 3/4" Wide adhesive line guarantees easy and reliable sealing
- Chevron with corner seals assures dust-free, sterile presentation
- Transparent side of pouches facilitates visual inspection of sterilisable materials and instruments
They have indicator imprints on both the inside and outside. This unique feature monitors the effective penetration of steam or ethylene oxide gas on the inside of the package. The hermetic seal will not open in steam autoclaves and is not affected by ethylene oxide gas.
These pouches save money by eliminating the need for separate indicator strips, the self-sealing feature eliminates the need for expensive and troublesome heat sealers and heat-sealing machines yet provides a seal as effective in maintaining sterility as any heat seal.