Magnesium oxide Ph. Eur., Macron Fine Chemicals™
Catalog # P119-33
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:40.3 g/mol
CAS Number:1309-48-4
Formula:MgO
MDL Number:MFCD00011109