Ethyl acetate NF, J.T.Baker®
Catalog # 9278-07
Supplier: Avantor
CAS Number:
Specifications
- Pk:20 L
- Seq:5302
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:99.0 - 100.5 %
- NF - Assay (CH₃COOC₂H₅):98.0 - 102.0 %
- NF - Identification:Passes Test
- NF - Specific Gravity at 25°/25°C:0.894 - 0.898
- NF - Acidity:Passes Test
- NF - Readily Carbonizable Substances:Passes Test
- NF - Nonvolatile Residue:≤0.02 %
- Chromatographic Purity: Acetaldehyde:≤0.1 %
- Chromatographic Purity: Ethyl isobutyl ether:≤0.10 %
- Chromatographic Purity: Methyl compounds:≤0.1 %
- Chromatographic Purity: Other impurities:≤0.30 %
Specification Test Results
About this item
:
:88.11 g/mol
UN:1173
ADR:3,II
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171