Anti-Ptprc Rat Monoclonal Antibody (PE) [clone: 30-F11]
Catalog # ABNOMAB15481
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase, receptor type C
- Antigen symbol:Ptprc
- Clonality:Monoclonal
- Clone:30-F11
- Conjugation:PE (Phycoerythrin)
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Form:Liquid
- Gene ID:19264
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light.
- Immunogen:Mouse thymocytes and splenocytes.
- Purification:Affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rat monoclonal antibody raised against native mouse Ptprc.
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: 30-F11
Conjugation: PE
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse