Oxygen depletion monitor and warning alarm system
478-0553
- Description:Cryogenic monitoring panel for use with monitoring O₂/temperature and valve status
- Seq:01
- Pk:1
Protects against O₂ deficiency where personnel are at risk from nitrogen leakage.
- Audible and visual alarms on all modules to provide low and critically low oxygen warning
- Base unit cannot be turned off, giving additional security to user
- Battery powered with 24 months continuous operation with separate low battery warnings
- No mains connection needed, simply screws to the wall
- Optional battery power repeater module to provide LCD display locally and/or remotely