Order Entry
Northern Ireland
ContactUsLinkComponent
Acid detergent descaler; RBS T 310
Acid detergent descaler; RBS T 310
Catalog # CHEPT310/E123
Supplier:  CHEMICAL
undefined
Acid detergent descaler; RBS T 310
Catalog # CHEPT310/E123
Supplier:  CHEMICAL

Specifications

  • Description:
    Acid detergent descaler; RBS T 310
  • Pk:
    5 L
  • pH range:
    2.2 (2%)

Specifications

About this item

Cleaning agent, descaler and oxides remover.

  • Low use concentration
  • High detergency power
  • Contains citric acid

RBS T 310 is suitable for manual cleaning, soaking and ultrasonic cleaning of items and laboratory equipment. It was especially designed to remove mineral residues such as calcareous deposits, rust, insoluble salts etc.

Ordering information: Available in box of 4 × 5 litres. Please contact Avantor for details.