Order Entry
Northern Ireland
ContactUsLinkComponent
Polyvalent detergent, RBS T 115
Polyvalent detergent, RBS T 115
Catalog # CHEPT115/E123
Supplier:  CHEMICAL
undefined
Polyvalent detergent, RBS T 115
Catalog # CHEPT115/E123
Supplier:  CHEMICAL

Specifications

  • Description:
    Polyvalent detergent, RBS T 115
  • Pk:
    5 L
  • pH range:
    11.7 (2%)

Specifications

About this item

For manual cleaning, soaking and ultrasonic cleaning of glassware and laboratory equipment.

  • Phosphate-free
  • Concentrated solution
  • High emulsifying and degreasing power

RBS T 115 is an alkaline cleaning agent recommended for manual cleaning, soaking and ultrasonic cleaning of glassware and laboratory equipment. It efficiently removes organic and biological residues, distilation residues etc. Particularly adapted for use in water analysis and environmental laboratories

Ordering information: Available in box of 4 × 5 litres. Please contact Avantor for details.
Caution: Not recommended for use on white metals.