ContactUsLinkComponent
Transformation kit, EZ-Yeast™, MP Biomedicals
Transformation kit, EZ-Yeast™, MP Biomedicals
Catalog # ICNA112100200
Supplier:  MP Biomedicals
CAS Number:  
Transformation kit, EZ-Yeast™, MP Biomedicals
Catalog # ICNA112100200
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    EZ-Yeast™ Transformation Kit
Specifications
About this item
The EZ-Yeast™ Transformation Kit is designed for high throughput transformation with efficiencies up to 10³ transformants per μg plasmid DNA. It is ideal for cases where a large number of transformations must be performed, but only a few transformants are needed. Perfect for a quick analysis of potentially positive two-hybrid colonies, it does not require fully competent cells prior to transformation.
Delivery: Kit contains EZ-Transformation Solution and carrier DNA.