VWR®, Calibrated Electronic Thermometers with Stainless Steel Probe
These high accuracy digital thermometers, with chemical resistant probes, monitor the temperature of pharmaceuticals, vaccines, chemicals, blood and blood products, food products and more.
- Features include: 10-event recording, max./min. temperature readings, timer and alarm
- Calibration points: 0; 100 °C
: Traceable to NIST, individually serialised, CE marked, ISO/IEC 17025:2005.
: Includes accredited calibration documentation and multi-language instructions (English, French, German, Italian, Portuguese and Spanish) and AAA battery.
Frequently Bought Together