Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ACCSL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-ACCSL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-9062R-FITC
Supplier:  Bioss
Anti-ACCSL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-9062R-FITC
Supplier:  Bioss

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    1-aminocyclopropane-1-carboxylate synthase homolog (Arabidopsis)(non-functional)-like
  • Antigen symbol:
    ACCSL
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    1A1L2_HUMAN|ACC synthase like protein 2|1 aminocyclopropane 1 carboxylate synthase like protein 2|ACC synthaseSL.|1-aminocyclopropane-1-carboxylate synthase-like protein 2|1 aminocyclopropane 1 carboxylate synthase homolog Arabidopsisnon functional like
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Belongs to the class-I pyridoxal-phosphate-dependent aminotransferase family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACCSL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat