Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:EF-hand domain (C-terminal) containing 2
- Antigen symbol:EFHC2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:80258
- Antigen synonyms:dJ1158H2.1|MRX74
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
EFHC2 contains 3 DM10 domains, followed by 3 putative EF-hand motifs. There are two named isoforms produced by alternative splicing. EFHC2 can be used as a quantitative trait locus for fear recognition in Turner syndrome.EF-HC2 is a 749 amino acid protein containing three DM10 domains and an EF-hand domain. EF-HC2 shares 41.6% homology with EF-HC1, and is widely expressed in peripheral tissues and central nervous system. The gene encoding EF-HC2 maps to human chromosome Xp11.3 and is critical for recognition of facial fear and harm avoidance. Turner syndrome, characterized by deficits in social cognition and recognition of facial fear, has been linked to the EF-HC2 gene. Deletion of the EF-HC2 gene may be associated with Norrie disease, an X-linked disorder that primarily effects the eye, and may also be linked to juvenile myoclonic epilepsy. EF-HC2 exists as two isoforms due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EFHC2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat