Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Dual Specificity Phosphatase 13
- Antigen symbol:DUSP13
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:51207
- Antigen synonyms:MDSP|DUSP13A|BEDP|SKRP4|DUSP13B|TMDP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Members of the protein-tyrosine phosphatase superfamilycooperate with protein kinases to regulate cell proliferation anddifferentiation. This superfamily is separated into two familiesbased on the substrate that is dephosphorylated. One family, thedual specificity phosphatases (DSPs) acts on both phosphotyrosineand phosphoserine/threonine residues. This gene encodes differentbut related DSP proteins through the use of non-overlapping openreading frames, alternate splicing, and presumed differenttranscription promoters. Expression of the distinct proteins fromthis gene has been found to be tissue specific and the proteins maybe involved in postnatal development of specific tissues. A proteinencoded by the upstream ORF was found in skeletal muscle, whereasthe encoded protein from the downstream ORF was found only intestis. In mouse, a similar pattern of expression was found.Multiple alternatively spliced transcript variants were described,but the full-length sequence of only some were determined.[provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DUSP13
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat