Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Protein Tyrosine Phosphatase, Non-Receptor Type 13
- Antigen symbol:PTPN13
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5783
- Antigen synonyms:PTP-E1|protein tyrosine phosphatase 1E|PTP-BL|hPTPE1|PTP1E|APO-1/CD95 (Fas)-associated phosphatase|PNP1|FAP-1|PTP-BAS|fas-associated protein-tyrosine phosphatase 1|DKFZp686J1497|tyrosine-protein phosphatase non-receptor type 13|protein-tyrosine phosphatase 1|nonreceptor type 13|protein-tyrosine phosphatase PTPL1|PTPLE|PTPL1|Fas-associated phosphatase-1|protein tyrosine phosphatase|protein-tyrosine phosphatase 1E|Fas-associated
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
PTPN13 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP is a large protein that possesses a PTP domain at C-terminus, and multiple noncatalytic domains, which include a domain with similarity to band 4.1 superfamily of cytoskeletal associated proteins, a region consisting of five PDZ domains, and a leucine zipper motif. This PTP was found to interact with, and dephosphorylate Fas receptor, as well as I-kappa-B-alpha through the PDZ domains, which suggested its role in Fas mediated programmed cell death. This PTP was also shown to interact with GTPase-activating protein, and thus may function as a regulator of Rho signaling pathway.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PTPN13
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat