Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Carnitine Palmitoyltransferase 1B
- Antigen symbol:CPT1B
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1375
- Antigen synonyms:CPT1B|FLJ58750|carnitine palmitoyltransferase 1B (muscle)|carnitine palmitoyltransferase 1B (muscle) |KIAA1670|CPTI|FLJ55729|carnitine palmitoyltransferase 1A (muscle)|carnitine O-palmitoyltransferase I|carnitine palmitoyltransferase I|M-CPT1|muscle|mitochondrial muscle isoform|carnitine palmitoyltransferase I-like|CPT1-M|carnitine palmitoyltransferase 1B|MCCPT1|CPT1M|MCPT1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:530-580/772
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and read-through transcripts are expressed from the upstream locus that include exons from this gene. [provided by RefSeq, Jun 2009].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CPT1B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity: