Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-IL18RAP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-IL18RAP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-2616R-A350
Supplier:  Bioss
Anti-IL18RAP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-2616R-A350
Supplier:  Bioss

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 18 Receptor Accessory Protein
  • Antigen symbol:
    IL18RAP
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8807
  • Antigen synonyms:
    IL-18Rbeta|IL-1RAcPL|CDw218b|CD218b|IL18RB|IL-1R-7|IL-1R7|IL-18R-beta|ACPL|IL-18RAcP
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    20-50/599
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Required for the high affinity binding of interleukin 18 (IL-18) to its receptor complex (By similarity). Together with IL18R1 mediates IL-18-dependent activation of NF-kappa-B and JNK.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL18RAP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat