Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 14
- Antigen symbol:TNFRSF14
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:8764
- Antigen synonyms:HVEA|Herpesvirus entry mediator|HVEM|CD270|ATAR|TR2|LIGHTR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
TNFRSF14 is a type I membrane protein belonging to the TNF receptor superfamily. This receptor mediates herpes virus entry into cells during infection. TNFRSF14 is able to inhibit the proliferation, activation, and cytokine production of T cells. It has an extracellular domain containing several cysteine-rich repeats and a short cytoplasmic region containing a TRAF (TNF receptor-associated factor) interaction domain. The extracellular domain of TNFRSF14 interacts with the herpes simplex virus envelope glycoprotein D. TNFRSF14 binds two cellular ligands: lymphotoxin alpha and LIGHT. LIGHT is a transmembrane protein expressed and shed from the surface of activated T cells, exhibits inducible expression, and competes with HSV glycoprotein D for HVEM, a receptor expressed by T lymphocytes. The LIGHT:TNFRSF14 interaction controls immune response functions by cell death induction as well as cell activation. TNFRSF14 is expressed by peripheral blood T cells, B cells, monocytes and in various tissues enriched in lymphoid cells.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: TNFRSF14
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human