Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Kh Domain Containing, Rna Binding, Signal Transduction Associated 1
- Antigen symbol:KHDRBS1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Sam 68 is a protein that is phosphorylated on tyrosine and functions as a substrate for Src family tyrosine kinases during mitosis. Sam 68 also associates with several SH2 and SH3 domain-containing signaling proteins, such as GRB2 and PLC ?. Originally cloned as Ras GAP-associated p62, further investigations have shown that Sam 68 and Ras GAP-associated p62 are not antigenically related, nor are they encoded by the same gene. Like Sam 68, the Sam 68-like mammalian proteins, SLM-1 and SLM-2, demonstrate RNA binding activity. Also like Sam 68, SLM-1 is tyrosine phosphorylated and functions as an adapter protein for signaling molecules, including GRB2, PLC ?, Fyn and Ras GAP. SLM-2 is not tyrosine phosphorylated, nor does it appear to associate with GRB2, PLC ?, Fyn or Ras GAP, indicating that SLM-2 may not be an adapter protein for these proteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KHDRBS1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat