To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Glycine Ceavage System Protein H (aminomethyl carrier)
- Antigen symbol:GCSH
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2653
- Antigen synonyms:NKH|GCE
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
GCSH is a 173 amino acid mitochondrial protein that contains one lipoyl-binding domain and belongs to the gcvH family. Defects in the gene encoding GCSH are the cause of glycine encephalopathy (GCE), an autosomal recessive disease that is also referred to as non-ketotic hyperglycinemia (NKH). Characterized by severe neurological symptoms, patients with GCE have a large amount of glycine accumulated in their body fluids. The gene encoding GCSH maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GCSH
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat